DL-Methioninmethylester Hydrochlorid

Lieferant: SIA ENAMINE

Synonyme: Methyl 2-amino-4-(methylthio)butanoate hydrochloride, H-DL-Met-OMe·HCl

EN30009315.1G
ENAMEN30009315.1GEA 0 EUR
ENAMEN30009315.1G
DL-Methioninmethylester Hydrochlorid
DL-Methioninmethylester Hydrochlorid
Formel: C₆H₁₃NO₂S·HCl
Molecular Weight: 199,7 g/mol
MDL: MFCD00067540
CAS-Nummer: 16118-36-8
EINECS: 240-285-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR