3-(Trifluormethyl)pyridin
Lieferant: SIA ENAMINE
EN30009350.1G
ENAMEN30009350.1GEA
0
EUR
ENAMEN30009350.1G
3-(Trifluormethyl)pyridin
3-(Trifluoromethyl)pyridine
Formel:
C₆H₄F₃N Siedepunkt: 110…113 °C (1013 hPa) |
MDL:
MFCD00100032 CAS-Nummer: 3796-23-4 EINECS: 223-268-2 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...