1-Chlor-4-(4-fluorphenyl)phthalazin

Lieferant: SIA ENAMINE

EN30009382.1G
ENAMEN30009382.1GEA 0 EUR
ENAMEN30009382.1G
1-Chlor-4-(4-fluorphenyl)phthalazin
1-Chloro-4-(4-fluorophenyl)phthalazine
Formel: C₁₄H₈ClFN₂
MDL: MFCD06253841
CAS-Nummer: 155937-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR