3,4-Dihydroxy-5-methoxybenzaldehyd

Lieferant: SIA ENAMINE

EN30009520.1G
ENAMEN30009520.1GEA 0 EUR
ENAMEN30009520.1G
3,4-Dihydroxy-5-methoxybenzaldehyd
3,4-Dihydroxy-5-methoxybenzaldehyde
Formel: C₈H₈O₄
MDL: MFCD00016610
CAS-Nummer: 3934-87-0
EINECS: 223-513-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR