5-Amino-3-(4-chlorphenyl)isoxazol

Lieferant: SIA ENAMINE

Synonyme: 3-(4-Chlorophenyl)isoxazol-5-amine

EN30009527.1G
ENAMEN30009527.1GEA 0 EUR
ENAMEN30009527.1G
5-Amino-3-(4-chlorphenyl)isoxazol
5-Amino-3-(4-chlorophenyl)isoxazole
Formel: C₉H₇ClN₂O
Molecular Weight: 194,6177 g/mol
MDL: MFCD01569450
CAS-Nummer: 33866-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR