3-Brom-5-ethoxy-4-hydroxybenzonitril

Lieferant: SIA ENAMINE

EN30009546.1G
ENAMEN30009546.1GEA 0 EUR
ENAMEN30009546.1G
3-Brom-5-ethoxy-4-hydroxybenzonitril
3-Brom-5-ethoxy-4-hydroxybenzonitril
Formel: C₉H₈BrNO₂
MDL: MFCD01123035
CAS-Nummer: 330462-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR