2-Amino-N-cyclopropylbenzamid
Lieferant: SIA ENAMINE
EN30009734.1G
ENAMEN30009734.1GEA
0
EUR
ENAMEN30009734.1G
2-Amino-N-cyclopropylbenzamid
2-Amino-N-cyclopropylbenzamid
Formel:
C₁₀H₁₂N₂O |
MDL:
MFCD03015446 CAS-Nummer: 30510-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...