4-Amino-N-methylbenzamid

Lieferant: SIA ENAMINE

EN30009745.1G
ENAMEN30009745.1GEA 0 EUR
ENAMEN30009745.1G
4-Amino-N-methylbenzamid
4-Amino-N-methylbenzamid
Formel: C₈H₁₀N₂O
MDL: MFCD00665792
CAS-Nummer: 6274-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR