2-Acetyl-5-brombenzofuran

Lieferant: SIA ENAMINE

Synonyme: 1-(5-Bromobenzofuran-2-yl)ethanone, 2-Acetyl-5-bromobenzo[b]furan

EN30009787.1G
ENAMEN30009787.1GEA 0 EUR
ENAMEN30009787.1G
2-Acetyl-5-brombenzofuran
2-Acetyl-5-brombenzofuran
Formel: C₁₀H₇BrO₂
Molecular Weight: 239,07 g/mol
MDL: MFCD00276737
CAS-Nummer: 38220-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR