2-Amino-N-propylbenzamid

Lieferant: SIA ENAMINE

EN30009840.1G
ENAMEN30009840.1GEA 0 EUR
ENAMEN30009840.1G
2-Amino-N-propylbenzamid
2-Amino-N-propylbenzamid
Formel: C₁₀H₁₄N₂O
MDL: MFCD02374350
CAS-Nummer: 56814-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR