1-((4-Methylphenyl)sulfonyl)-4-piperidincarbonsäurehydrazid

Lieferant: SIA ENAMINE

EN30009928.1G
ENAMEN30009928.1GEA 0 EUR
ENAMEN30009928.1G
1-((4-Methylphenyl)sulfonyl)-4-piperidincarbonsäurehydrazid
1-((4-Methylphenyl)sulfonyl)-4-piperidincarbonsäurehydrazid
Formel: C₁₃H₁₉N₃O₃S
MDL: MFCD00759493
CAS-Nummer: 324531-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR