Methyl-3-(brommethyl)benzoat

Lieferant: SIA ENAMINE

Synonyme: Methyl 3-(bromomethyl)benzoate

EN30010007.1G
ENAMEN30010007.1GEA 0 EUR
ENAMEN30010007.1G
Methyl-3-(brommethyl)benzoat
Methyl-3-(brommethyl)benzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229,07 g/mol
MDL: MFCD00051437
CAS-Nummer: 1129-28-8
EINECS: 419-100-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR