4-(1-Aminoethyl)pyridin

Lieferant: SIA ENAMINE

Synonyme: 1-(4-Pyridyl)ethylamin, 1-Pyridin-4-ylethylamin

EN30010016.1G
ENAMEN30010016.1GEA 0 EUR
ENAMEN30010016.1G
4-(1-Aminoethyl)pyridin
4-(1-Aminoethyl)pyridine
Formel: C₇H₁₀N₂
Molecular Weight: 122,17 g/mol
MDL: MFCD02245004
CAS-Nummer: 50392-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR