Methyl-3,3-dimethoxypropionat

Lieferant: SIA ENAMINE

Synonyme: Methyl 3,3-dimethoxypropanoate, Methyl 3,3-dimethoxypropionate

EN300100162.1G
ENAMEN300100162.1GEA 0 EUR
ENAMEN300100162.1G
Methyl-3,3-dimethoxypropionat
Methyl-3,3-dimethoxypropionat
Formel: C₆H₁₂O₄
Molecular Weight: 148,16 g/mol
MDL: MFCD00010650
CAS-Nummer: 7424-91-1
EINECS: 231-055-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR