3-Chlorthiophen
Lieferant: SIA ENAMINE
EN300100252.1G
ENAMEN300100252.1GEA
0
EUR
ENAMEN300100252.1G
3-Chlorthiophen
3-Chlorthiophen
Formel:
C₄H₃ClS Molecular Weight: 118,59 g/mol Siedepunkt: 133…135 °C (1013 hPa) Dichte: 1,276 g/cm³ (20 °C) |
MDL:
MFCD00043887 CAS-Nummer: 17249-80-8 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...