2-Chlor-3-(chlormethyl)-7-methoxychinolin

Lieferant: SIA ENAMINE

EN30010029.1G
ENAMEN30010029.1GEA 0 EUR
ENAMEN30010029.1G
2-Chlor-3-(chlormethyl)-7-methoxychinolin
2-Chloro-3-(chloromethyl)-7-methoxyquinoline
Formel: C₁₁H₉Cl₂NO
CAS-Nummer: 73863-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR