3-Amino-6-fluor-1H-indazol

Lieferant: SIA ENAMINE

EN300100294.1G
ENAMEN300100294.1GEA 0 EUR
ENAMEN300100294.1G
3-Amino-6-fluor-1H-indazol
3-Amino-6-fluoro-1H-indazole
Formel: C₇H₆FN₃
MDL: MFCD06739129
CAS-Nummer: 404827-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR