3-Fluorthiophenol
Lieferant: SIA ENAMINE
Synonyme:
3-Fluorphenylmercaptan, 3-Fluorbenzolthiol, m-Fluorthiophenol, 1-Fluor-3-mercaptobenzol, 3-Fluorbenzol-1-thiol
EN300100319.1G
ENAMEN300100319.1GEA
0
EUR
ENAMEN300100319.1G
3-Fluorthiophenol
3-Fluorthiophenol
Formel:
C₆H₅FS Molecular Weight: 128,17 g/mol Siedepunkt: 164…166 °C (1013 hPa) Dichte: 1,18 g/cm³ (30 °C) |
MDL:
MFCD00040227 CAS-Nummer: 2557-77-9 EINECS: 219-876-2 UN: 3071 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...