3,5-Dimethyl-1H-pyrazol-1-methanol

Lieferant: SIA ENAMINE

Synonyme: 3,5-Dimethyl-1-(hydroxymethyl)-1H-pyrazole

EN300100417.1G
ENAMEN300100417.1GEA 0 EUR
ENAMEN300100417.1G
3,5-Dimethyl-1H-pyrazol-1-methanol
3,5-Dimethyl-1H-pyrazol-1-methanol
Formel: C₆H₁₀N₂O
MDL: MFCD00020738
CAS-Nummer: 85264-33-1
EINECS: 286-553-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR