2-Brom-6-fluoranilin
Lieferant: SIA ENAMINE
|
Achtung
|
EN300100611.1G
ENAMEN300100611.1GEA
0
EUR
ENAMEN300100611.1G
2-Brom-6-fluoranilin
2-Brom-6-fluoranilin
Formel:
C₆H₅BrFN Molecular Weight: 190,01 g/mol |
MDL:
MFCD01310982 CAS-Nummer: 65896-11-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...