(2,4-Difluorphenyl)essigsäure

Lieferant: SIA ENAMINE
Achtung

Synonyme: 2,4-Difluorophenylacetic acid

EN300100639.1G
ENAMEN300100639.1GEA 0 EUR
ENAMEN300100639.1G
(2,4-Difluorphenyl)essigsäure
(2,4-Difluorphenyl)essigsäure
Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172,13 g/mol
Siedepunkt: 259 °C (1013 hPa)
Schmelzpunkt: 117…119 °C
MDL: MFCD00009999
CAS-Nummer: 81228-09-3
EINECS: 279-709-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR