(3,4-Difluorphenyl)essigsäure

Lieferant: SIA ENAMINE
Achtung

Synonyme: 3,4-Difluorophenylacetic acid

EN300100640.1G
ENAMEN300100640.1GEA 0 EUR
ENAMEN300100640.1G
(3,4-Difluorphenyl)essigsäure
(3,4-Difluorphenyl)essigsäure
Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172,13 g/mol
Siedepunkt: 259 °C (1013 hPa)
Schmelzpunkt: 40…42 °C
MDL: MFCD00010002
CAS-Nummer: 658-93-5
EINECS: 211-527-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR