(2,3-Difluorphenyl)essigsäure

Lieferant: SIA ENAMINE
Achtung

Synonyme: 2-(2,3-Difluorophenyl)acetic acid

EN300100654.1G
ENAMEN300100654.1GEA 0 EUR
ENAMEN300100654.1G
(2,3-Difluorphenyl)essigsäure
(2,3-Difluorphenyl)essigsäure
Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172,13 g/mol
MDL: MFCD00040968
CAS-Nummer: 145689-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR