(3-(Trifluormethoxy)phenyl)essigsäure

Lieferant: SIA ENAMINE
Achtung

Synonyme: 3-(Trifluoromethoxy)phenylacetic acid

EN300100655.1G
ENAMEN300100655.1GEA 0 EUR
ENAMEN300100655.1G
(3-(Trifluormethoxy)phenyl)essigsäure
(3-(Trifluormethoxy)phenyl)essigsäure
Formel: (C₆H₄(OCF₃))CH₂COOH
Molecular Weight: 220,15 g/mol
MDL: MFCD00082480
CAS-Nummer: 203302-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR