Isochinolin-4-carbaldehyd

Lieferant: SIA ENAMINE

EN300100667.1G
ENAMEN300100667.1GEA 0 EUR
ENAMEN300100667.1G
Isochinolin-4-carbaldehyd
Isochinolin-4-carbaldehyd
Formel: C₁₀H₇NO
MDL: MFCD00829440
CAS-Nummer: 22960-16-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR