Benzyl-(S)-(+)-glycidylether

Lieferant: SIA ENAMINE

EN300100690.1G
ENAMEN300100690.1GEA 0 EUR
ENAMEN300100690.1G
Benzyl-(S)-(+)-glycidylether
Benzyl-(S)-(+)-glycidylether
Formel: C₁₀H₁₂O₂
MDL: MFCD00054428
CAS-Nummer: 16495-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR