1,2,4-Triazol-3-carbonsäure

Lieferant: SIA ENAMINE

Synonyme: 1H-1,2,4-Triazole-3-carboxylic acid

EN300100741.1G
ENAMEN300100741.1GEA 0 EUR
ENAMEN300100741.1G
1,2,4-Triazol-3-carbonsäure
1,2,4-Triazole-3-carboxylic acid
Formel: C₃H₃N₃O₂
Molecular Weight: 113,0748 g/mol
MDL: MFCD00078268
CAS-Nummer: 4928-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR