3-(Methylsulfonyl)benzoesäure

Lieferant: SIA ENAMINE

EN30010083.1G
ENAMEN30010083.1GEA 0 EUR
ENAMEN30010083.1G
3-(Methylsulfonyl)benzoesäure
3-(Methylsulphonyl)benzoic acid
Formel: C₈H₈O₄S
Molecular Weight: 200,21 g/mol
MDL: MFCD00216488
CAS-Nummer: 5345-27-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR