Dimethylcyclopropan-1,1-dicarboxylat

Lieferant: SIA ENAMINE

Synonyme: Dimethyl 1,1-cyclopropanedicarboxylate, 1,1-Cyclopropanedicarboxylic acid dimethyl ester

EN300100932.1G
ENAMEN300100932.1GEA 0 EUR
ENAMEN300100932.1G
Dimethylcyclopropan-1,1-dicarboxylat
Dimethylcyclopropan-1,1-dicarboxylat
Formel: C₇H₁₀O₄
Molecular Weight: 158,15 g/mol
MDL: MFCD00192079
CAS-Nummer: 6914-71-2
EINECS: 414-240-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR