Indol-6-carbonitril

Lieferant: SIA ENAMINE

Synonyme: 6-Indolcarbonitril, 6-Cyanindol, 6-Cyan-1H-indol, 1H-Indol-6-carbonitril

EN300100999.1G
ENAMEN300100999.1GEA 0 EUR
ENAMEN300100999.1G
Indol-6-carbonitril
Indol-6-carbonitril
Formel: C₉H₆N₂
Molecular Weight: 142,16 g/mol
MDL: MFCD00016732
CAS-Nummer: 15861-36-6
EINECS: 239-988-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR