3-(2-Carbamoylphenoxy)propionsäure

Lieferant: SIA ENAMINE

EN300101066.1G
ENAMEN300101066.1GEA 0 EUR
ENAMEN300101066.1G
3-(2-Carbamoylphenoxy)propionsäure
3-(2-Carbamoylphenoxy)propionsäure
Formel: C₁₀H₁₁NO₄
MDL: MFCD02364319
CAS-Nummer: 103204-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR