2,6-Difluor-3-nitrobenzoesäure
Lieferant: SIA ENAMINE
EN300101239.1G
ENAMEN300101239.1GEA
0
EUR
ENAMEN300101239.1G
2,6-Difluor-3-nitrobenzoesäure
2,6-Difluoro-3-nitrobenzoic acid
Formel:
C₇H₃F₂NO₄ |
MDL:
MFCD00119291 CAS-Nummer: 83141-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...