Methyl-2,2-difluor-2-(fluorsulfonyl)acetat
Lieferant: SIA ENAMINE
EN300101267.1G
ENAMEN300101267.1GEA
0
EUR
ENAMEN300101267.1G
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat
Formel:
C₃H₃F₃O₄S Molecular Weight: 192,11 g/mol |
MDL:
MFCD00144316 CAS-Nummer: 680-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...