3-(3-Fluor-4-methoxybenzoyl)propionsäure

Lieferant: SIA ENAMINE

EN300101432.1G
ENAMEN300101432.1GEA 0 EUR
ENAMEN300101432.1G
3-(3-Fluor-4-methoxybenzoyl)propionsäure
3-(3-Fluor-4-methoxybenzoyl)propionsäure
Formel: C₁₁H₁₁FO₄
Molecular Weight: 226,20 g/mol
MDL: MFCD00020538
CAS-Nummer: 347-63-7
EINECS: 206-474-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR