1-(2,4-Dimethylphenyl)-2-harnstoff

Lieferant: SIA ENAMINE

Synonyme: 2,4-Dimethylphenylurea

EN30010145.1G
ENAMEN30010145.1GEA 0 EUR
ENAMEN30010145.1G
1-(2,4-Dimethylphenyl)-2-harnstoff
1-(2,4-Dimethylphenyl)-2-harnstoff
Formel: C₉H₁₂N₂O
MDL: MFCD00025409
CAS-Nummer: 2990-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR