α,α,α,2-Tetrafluor-m-toluylsäure

Lieferant: SIA ENAMINE

EN300101462.1G
ENAMEN300101462.1GEA 0 EUR
ENAMEN300101462.1G
α,α,α,2-Tetrafluor-m-toluylsäure
α,α,α,2-Tetrafluor-m-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208,11 g/mol
MDL: MFCD00040980
CAS-Nummer: 115029-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR