Methyl-5-methylnicotinat

Lieferant: SIA ENAMINE

Synonyme: Methyl 5-methylnicotinate

EN300101528.1G
ENAMEN300101528.1GEA 0 EUR
ENAMEN300101528.1G
Methyl-5-methylnicotinat
Methyl-5-methylnicotinat
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD01075739
CAS-Nummer: 29681-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR