5-Methylnicotinamid

Lieferant: SIA ENAMINE

EN300101531.1G
ENAMEN300101531.1GEA 0 EUR
ENAMEN300101531.1G
5-Methylnicotinamid
5-Methylnicotinamid
Formel: C₇H₈N₂O
MDL: MFCD05664041
CAS-Nummer: 70-57-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR