1-Brom-3,5-dimethoxybenzol

Lieferant: SIA ENAMINE

EN300101669.1G
ENAMEN300101669.1GEA 0 EUR
ENAMEN300101669.1G
1-Brom-3,5-dimethoxybenzol
1-Brom-3,5-dimethoxybenzol
Formel: C₈H₉BrO₂
MDL: MFCD06200685
CAS-Nummer: 20469-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR