2-Chlor-5-fluorpyrimidin

Lieferant: SIA ENAMINE

EN300101688.1G
ENAMEN300101688.1GEA 0 EUR
ENAMEN300101688.1G
2-Chlor-5-fluorpyrimidin
2-Chloro-5-fluoropyrimidine
Formel: C₄H₂ClFN₂
Molecular Weight: 132,52 g/mol
MDL: MFCD03788197
CAS-Nummer: 62802-42-0
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR