5-Amino-3-(4-fluorphenyl)isoxazol

Lieferant: SIA ENAMINE

EN30010186.1G
ENAMEN30010186.1GEA 0 EUR
ENAMEN30010186.1G
5-Amino-3-(4-fluorphenyl)isoxazol
5-Amino-3-(4-fluorophenyl)isoxazole
Formel: C₉H₇FN₂O
MDL: MFCD03407398
CAS-Nummer: 81465-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR