6-Amino-1-naphthalinsulfonsäure

Lieferant: SIA ENAMINE

Synonyme: 6-Amino-1-naphthalenesulphonic acid

EN300101916.1G
ENAMEN300101916.1GEA 0 EUR
ENAMEN300101916.1G
6-Amino-1-naphthalinsulfonsäure
6-Amino-1-naphthalenesulphonic acid
Formel: C₁₀H₉NO₃S
Molecular Weight: 223,2500 g/mol
MDL: MFCD00035724
CAS-Nummer: 81-05-0
EINECS: 201-318-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR