Methyl-2,3,4-trifluorbenzoat

Lieferant: SIA ENAMINE

EN300101993.1G
ENAMEN300101993.1GEA 0 EUR
ENAMEN300101993.1G
Methyl-2,3,4-trifluorbenzoat
Methyl-2,3,4-trifluorbenzoat
Formel: C₈H₅F₃O₂
MDL: MFCD06201866
CAS-Nummer: 773873-68-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR