2-Brom-3-fluorpyridin

Lieferant: SIA ENAMINE

EN300102017.1G
ENAMEN300102017.1GEA 0 EUR
ENAMEN300102017.1G
2-Brom-3-fluorpyridin
2-Bromo-3-fluoropyridine
Formel: C₅H₃BrFN
MDL: MFCD04114128
CAS-Nummer: 40273-45-8
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR