2-Nitro-p-xylol
Lieferant: SIA ENAMINE
Synonyme:
2,5-Dimethylnitrobenzene, Nitro-p-xylene, 1,4-Dimethyl-2-nitrobenzene
EN300102019.1G
ENAMEN300102019.1GEA
0
EUR
ENAMEN300102019.1G
2-Nitro-p-xylol
2-Nitro-p-xylol
Formel:
C₈H₉NO₂ Molecular Weight: 151,16 g/mol Siedepunkt: 240 °C (1013 hPa) Schmelzpunkt: –25 °C Dichte: 1,132 g/cm³ (20 °C) |
MDL:
MFCD00024284 CAS-Nummer: 89-58-7 EINECS: 201-920-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...