2-Cyan-4-fluoranilin

Lieferant: SIA ENAMINE

EN300102377.1G
ENAMEN300102377.1GEA 0 EUR
ENAMEN300102377.1G
2-Cyan-4-fluoranilin
2-Cyan-4-fluoranilin
Formel: C₇H₅FN₂
Molecular Weight: 136.13 g/mol
MDL: MFCD02683869
CAS-Nummer: 61272-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR