3-Amino-2-brom-6-methylpyridin

Lieferant: SIA ENAMINE

Synonyme: 2-Bromo-6-methylpyridin-3-amine, 3-Amino-2-bromo-6-picoline, 5-Amino-6-bromo-2-picoline

EN300102380.1G
ENAMEN300102380.1GEA 0 EUR
ENAMEN300102380.1G
3-Amino-2-brom-6-methylpyridin
3-Amino-2-bromo-6-methylpyridine
Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
MDL: MFCD03095078
CAS-Nummer: 126325-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR