4-Acetyl-2-chlorpyridin

Lieferant: SIA ENAMINE

Synonyme: 1-(2-chloropyridin-4-yl)ethan-1-one, 1-(2-chloropyridine-4-yl)ethanone

EN300102440.1G
ENAMEN300102440.1GEA 0 EUR
ENAMEN300102440.1G
4-Acetyl-2-chlorpyridin
4-Acetyl-2-chlorpyridin
Formel: C₇H₆ClNO
Molecular Weight: 155,58 g/mol
MDL: MFCD07699406
CAS-Nummer: 23794-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR