4-Acetamidobenzylalkohol

Lieferant: SIA ENAMINE

Synonyme: N-[4-(Hydroxymethyl)phenyl]acetamide, 4-Acetamidobenzyl alcohol, N-(4-(Hydroxymethyl)phenyl)acetamide

EN300102492.1G
ENAMEN300102492.1GEA 0 EUR
ENAMEN300102492.1G
4-Acetamidobenzylalkohol
4-Acetamidobenzylalkohol
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
MDL: MFCD00016868
CAS-Nummer: 16375-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR