2-Brom-4-fluorbenzaldehyd
Lieferant: SIA ENAMINE
EN300102567.1G
ENAMEN300102567.1GEA
0
EUR
ENAMEN300102567.1G
2-Brom-4-fluorbenzaldehyd
2-Bromo-4-fluorobenzaldehyde
Formel:
C₇H₄BrFO |
MDL:
MFCD00672923 CAS-Nummer: 59142-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...